Learn More
2-(Carboxymethyldisulfanyl)acetic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(Carboxymethyldisulfanyl)acetic Acid |
| CAS | 505-73-7 |
| Molecular Formula | C4H6O4S2 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C4H6O4S2/c5-3(6)1-9-10-2-4(7)8/h1-2H2,(H,5,6)(H,7,8) |
| SMILES | C(C(=O)O)SSCC(=O)O |
| IUPAC Name | 2-(carboxymethyldisulfanyl)acetic acid |
| Molecular Weight (g/mol) | 182.21 |
| Formula Weight | 181.9708 |