Learn More
2-Butyne-1,4-diol-(1,1,4,4)-d4, Diacetate, TRC
Spécification
Spécification
| Chemical Name or Material | 2-Butyne-1,4-diol-(1,1,4,4)-d4, Diacetate |
| CAS | 1020719-23-6 |
| Molecular Formula | C8H6D4O4 |
| Quantity | 10 mg |
| Synonym | 1,4-Diacetoxy-2-butyne-d4, NSC 76565-d4 |
| InChI Formula | InChI=1S/C8H10O4/c1-7(9)11-5-3-4-6-12-8(2)10/h5-6H2,1-2H3/i5D2,6D2 |
| SMILES | CC(OC([2H])([2H])C#CC([2H])([2H])OC(C)=O)=O |
| IUPAC Name | but-2-yne-1,4-diyl-d4 diacetate |
| Molecular Weight (g/mol) | 174.19 |
| Formula Weight | 174.08 |