Learn More
2-Bromotoluene-d7, CDN
Product Code. 30310072
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.50g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30310072
Brand: CDN CDND64040.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Bromotoluene-D7
CAS 1185306-95-9
Molecular Formula C7 D7 Br
Quantity 0.5 g
Synonym 5-Bromo-6-(methyl-d3)benzene-1,2,3,4-d4, 2-Bromotoluene-d7, 2-Bromotoluene-D7, 2-Bromotoluene-α,α,α,3,4,5,6-d7
InChI Formula InChI=1 S/C7H7Br/c1-6-4-2-3-5-7(6)8/h2-5 H,1H3/i1D3,2 D,3 D,4 D,5 D
SMILES [2 H]c1c([2 H])c([2 H])c(c(Br)c1[2 H])C([2 H])([2 H])[2 H]
IUPAC Name 1-bromo-2,3,4,5-tetradeuterio-6-(trideuteriomethyl)benzene
Molecular Weight (g/mol) 178.0776
Formula Weight 177.0171 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity