missing translation for 'onlineSavingsMsg'
Learn More

2-bromopyridine-5-boronic acid, 97%, Maybridge

Product Code. 10431694
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10431694

Brand: Thermo Scientific Maybridge MAY00056DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Specifications

CAS 223463-14-7
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C5H5BBrNO2
MDL Number MFCD03411558
Quantity 1 g
Synonym 2-bromopyridine-5-boronic acid, 6-bromopyridin-3-yl boronic acid, 6-bromopyridine-3-boronic acid, 2-bromo-5-pyridineboronic acid, 6-bromopyridin-3-lboronic acid, 2-bromo-5-pyridylboronic acid, 2-bromopyridyl-5-boronic acid, 6-bromo-3-pyridylboronic acid, 6-bromopyridin-3-yl-3-boronic acid, 6-bromo-3-pyridinylboronic acid
InChI Key BCYWDUVHAPHGIP-UHFFFAOYSA-N
SMILES OB(O)C1=CN=C(Br)C=C1
IUPAC Name (6-bromopyridin-3-yl)boronic acid
Molecular Weight (g/mol) 201.81
PubChem CID 2762706
Formula Weight 201.81
Percent Purity 97%
Chemical Name or Material 2-Bromopyridine-5-boronic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.