Learn More
2-Bromopyridine-4-carboxylic acid, 97%
Product Code. 11309855
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11309855
Brand: Thermo Scientific Alfa Aesar L19855.03
Request Bulk or Custom Format
This item requires a price update prior to being ordered.
Request a Price for this item here.
CAS: 66572-56-3 | C6H4BrNO2 | 202.007 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 66572-56-3
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.007
MDL Number MFCD01646069
InChI Key YBTKGKVQEXAYEM-UHFFFAOYSA-N
Synonym 2-bromoisonicotinic acid, 2-bromo-4-pyridinecarboxylic acid, 2-bromo-isonicotinic acid, 2-bromo-4-pyridine carboxylic acid, 4-pyridinecarboxylic acid, 2-bromo, 2-bromoisonicotic acid, 2-bromisonicotins, 2-bromopyridine-4-carboxylicacid, pubchem9211, acmc-209nvt
PubChem CID 222701
IUPAC Name 2-bromopyridine-4-carboxylic acid
SMILES C1=CN=C(C=C1C(=O)O)Br
Specifications
Melting Point ∽225°C (decomposition)
Quantity 1 g
Beilstein 471895
Formula Weight 202.01
Percent Purity 97%
Chemical Name or Material 2-Bromopyridine-4-carboxylic acid