Learn More
2-Bromopalmitic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-bromohexadecanoic acid |
| CAS | 18263-25-7 |
| Molecular Formula | C16 H31 Br O2 |
| Quantity | 1 g |
| Synonym | 2-Bromopalmitic Acid, 2-Bromohexadecanoic Acid, DL-2-Bromopalmitate, NSC 58378, alpha-Bromopalmitic Acid, 1-Bromopentadecanecarboxylic Acid, 2-Bromo-n-hexadecanoic Acid |
| InChI Formula | InChI=1S/C16H31BrO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15(17)16(18)19/h15H,2-14H2,1H3,(H,18,19) |
| SMILES | CCCCCCCCCCCCCCC(Br)C(=O)O |
| IUPAC Name | 2-bromohexadecanoic acid |
| Molecular Weight (g/mol) | 335.32 |
| Formula Weight | 334.151 |