missing translation for 'onlineSavingsMsg'
Learn More

2-Bromomethyl-1,3-dioxolane, 97%

Product Code. 11469566
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. Quantity unitSize
11469566 25 g 25g
11479566 100 g 100g
11489566 500 g 500g
3 options
This item is not returnable. View return policy

Product Code. 11469566

Brand: Thermo Scientific Alfa Aesar A19312.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4360-63-8 | C4H7BrO2 | 167.002 g/mol

2-Bromomethyl-1,3-dioxolane is used in the preparation of alfa, beta-unsaturated aldehyde and 1, 4-two aldehyde monoacetals synthesis of monomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromomethyl-1,3-dioxolane is used in the preparation of alfa, beta-unsaturated aldehyde and 1, 4-two aldehyde monoacetals synthesis of monomers.

Solubility
Immiscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4360-63-8
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.002
MDL Number MFCD00003214
InChI Key CKIIJIDEWWXQEA-UHFFFAOYSA-N
Synonym 2-bromomethyl-1,3-dioxolane, bromoacetaldehyde ethylene acetal, 1,3-dioxolane, 2-bromomethyl, 1,3-dioxolan-2-yl methyl bromide, bromomethyl dioxolane, acmc-1afrf, 2-bromomethyl-1,3-dioxolan, ksc495q7b, 2-bromomethyl-1,3 dioxolane, 2-bromomethyl-1, 3-dioxolane
PubChem CID 78068
IUPAC Name 2-(bromomethyl)-1,3-dioxolane
SMILES C1COC(O1)CBr

Specifications

Density 1.628
Boiling Point 175°C
Flash Point 62°C (145°F)
Refractive Index 1.482
Quantity 25 g
Beilstein 103309
Sensitivity Moisture sensitive
Solubility Information Immiscible with water.
Formula Weight 167.01
Percent Purity 97%
Chemical Name or Material 2-Bromomethyl-1,3-dioxolane
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.