Learn More
2-Bromofluorobenzene-d4, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1-Bromo-2-fluorobenzene-D8 |
| CAS | 50592-35-3 |
| Molecular Formula | C6 D4 Br F |
| Quantity | 0.5 g |
| Synonym | Benzene-1,2,3,4-d4, 5-bromo-6-fluoro- (9 CI), o-Bromofluorobenzene-d4, 5-Bromo-6-fluorobenzene-1,2,3,4-d4, 1-Bromo-2-fluorobenzene-d4, 1-Bromo-2-fluorobenzene-D4 |
| InChI Formula | InChI=1 S/C6H4BrF/c7-5-3-1-2-4-6(5)8/h1-4 H/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(Br)c(F)c1[2 H] |
| IUPAC Name | 1-bromo-2,3,4,5-tetradeuterio-6-fluorobenzene |
| Molecular Weight (g/mol) | 179.023 |
| Formula Weight | 177.9731 g/mol |
| Show More |