missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethyl ethyl ether, 90%

Product Code. p-3777635 Shop All Thermo Scientific Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10304502

Brand: Thermo Scientific 367400250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 592-55-2 | C4H9BrO | 153.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-55-2
Molecular Formula C4H9BrO
Molecular Weight (g/mol) 153.02
MDL Number MFCD00000237
InChI Key MMYKTRPLXXWLBC-UHFFFAOYSA-N
Synonym 2-bromoethyl ethyl ether, 2-ethoxyethyl bromide, 2-bromoethoxyethane, ethane, 1-bromo-2-ethoxy, 2-bromodiethyl ether, 1-ethoxy-2-bromoethane, 1-bromo-2-ethoxy-ethane, ether, 2-bromoethyl ethyl, 2-bromoethylethylether, 1-bromo-3-oxapentane
PubChem CID 61141
IUPAC Name 1-bromo-2-ethoxyethane
SMILES CCOCCBr

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Colorless to Brown
Density 1.3570g/mL
Boiling Point 125°C to 127°C
Flash Point 21°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2CH2OC2H5
Refractive Index 1.4435 to 1.4515
Quantity 25 g
Beilstein 01,338
Specific Gravity 1.357
Solubility Information Solubility in water: slightly soluble in water. Other solubilities: miscible in ethanol,ethyl ether
Formula Weight 153.02
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 2-Bromoethyl ethyl ether
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromoethyl ethyl ether
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.