Learn More
(2-Bromoethyl)benzene-d5, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | (2-Bromoethyl)benzene-d5 |
| CAS | 35845-64-8 |
| Molecular Formula | C8 D5 H4 Br |
| Quantity | 1 g |
| Synonym | Benzene-d5, (2-bromoethyl)- (9 CI), 1-(2-Bromoethyl)-2,3,4,5,6-pentadeuteriobenzene, 2-(Pentadeuterophenyl)ethyl bromide, (2-Bromoethyl)benzene-2,3,4,5,6-d5, (2-Bromoethyl)benzene-D5, Phenethylbromide-d5 (phenyl-d5) |
| InChI Formula | InChI=1 S/C8H9Br/c9-7-6-8-4-2-1-3-5-8/h1-5 H,6-7H2/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(CCBr)c([2 H])c1[2 H] |
| IUPAC Name | 1-(2-bromoethyl)-2,3,4,5,6-pentadeuteriobenzene |
| Molecular Weight (g/mol) | 190.092 |
| Formula Weight | 189.02 g/mol |
| Show More |