Learn More
2-Bromodibenzothiophene, 98%
Product Code. p-7041694
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 15461929
Brand: Thermo Scientific Alfa Aesar H64645.03
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
CAS: 22439-61-8 | C12H7BrS | 263.152 g/mol
Used as intermediate for organic light-emitting diode(OLED) materials and pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as intermediate for organic light-emitting diode(OLED) materials and pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 22439-61-8
Molecular Formula C12H7BrS
Molecular Weight (g/mol) 263.152
MDL Number MFCD00089285
InChI Key IJICRIUYZZESMW-UHFFFAOYSA-N
Synonym 2-bromodibenzo b,d thiophene, dibenzothiophene, 2-bromo, 2-bromobenzo b benzo b thiophene, 4-bromo-8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, acmc-209fwi, 2-bromo-dibenzothiophene, dibenzothiophene,2-bromo, 2-bromodibenzo b thiophene, 2-bromo-dibenzo b,d thiophene, dibenzo b,d thiophene, 2-bromo
PubChem CID 299508
IUPAC Name 2-bromodibenzothiophene
SMILES C1=CC=C2C(=C1)C3=C(S2)C=CC(=C3)Br
Specifications
Melting Point 123°C to 129°C (literature)
Quantity 1 g
Beilstein 142203
Solubility Information Slightly soluble in water.
Formula Weight 263.15
Percent Purity 98%
Chemical Name or Material 2-Bromodibenzothiophene