missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobutyric acid, 98%

Product Code. 11420357
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
This item is not returnable. View return policy

Product Code. 11420357

Brand: Thermo Scientific Alfa Aesar B21743.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 80-58-0 | C4H7BrO2 | 167.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-58-0
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.00
MDL Number MFCD00004216,MFCD00210111,MFCD00210112
InChI Key YAQLSKVCTLCIIE-UHFFFAOYNA-N
Synonym 2-bromobutyric acid, alpha-bromobutyric acid, butanoic acid, 2-bromo, butyric acid, 2-bromo, .alpha.-bromobytyric acid, butyric acid, .alpha.-bromo, dl-2-bromobutyric acid, a-bromobutyric acid, butyric acid, alpha-bromo, bromobutyric acid
PubChem CID 6655
IUPAC Name 2-bromobutanoic acid
SMILES CCC(Br)C(O)=O

Specifications

Melting Point -4°C
Density 1.567
Boiling Point 99°C to 103°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.472
Quantity 1000 g
UN Number UN3265
Beilstein 1720950
Merck Index 14,1413
Formula Weight 167.01
Percent Purity 98%
Chemical Name or Material 2-Bromobutyric acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.