missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobutane-d9, TRC

Product Code. 30338738
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30338738 50 mg 50mg
30334738 1 g 1g
30336999 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30338738 Supplier Toronto Research Chemicals Supplier No. TRCB68183750MG
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Bromobutane-d9
CAS 202392-72-1
Molecular Formula C4D9Br
Quantity 50 mg
Synonym (±)-2-Bromobutane-d9, (±)-sec-Butyl bromide-d9, 2-Bromobutane-d9, 2-Butyl bromide-d9, Methylethylbromomethane-d9, NSC 8417-d9, sec-Butyl bromide-d9
InChI Formula InChI=1S/C4H9Br/c1-3-4(2)5/h4H,3H2,1-2H3/i1D3,2D3,3D2,4D
SMILES BrC(C([2H])([2H])C([2H])([2H])[2H])([2H])C([2H])([2H])[2H]
IUPAC Name 2-bromo-1,1,1,2,3,3,4,4,4-nonadeuteriobutane
Molecular Weight (g/mol) 146.07
Formula Weight 145.05
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.