missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-chlorobenzoic acid, 98+%

Product Code. 11479332
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11479332

Brand: Thermo Scientific Alfa Aesar A10103.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21739-93-5 | C7H4BrClO2 | 235.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21739-93-5
Molecular Formula C7H4BrClO2
Molecular Weight (g/mol) 235.46
MDL Number MFCD00013982
InChI Key RBCPJQQJBAQSOU-UHFFFAOYSA-N
Synonym 6-bromo-3-chlorobenzoic acid, benzoic acid, 2-bromo-5-chloro, 2-bromo-5-chloro-benzoic acid, 5-chloro-2-bromobenzoic acid, pubchem3584, acmc-1co7w, 2-bromo-5-chlorobenzoicacid, ksc204k2j, benzoicacid, 2-bromo-5-chloro, zerenex e/9071908
PubChem CID 89027
IUPAC Name 2-bromo-5-chlorobenzoic acid
SMILES OC(=O)C1=CC(Cl)=CC=C1Br

Specifications

Melting Point 152°C to 156°C
Odor Odorless
Quantity 5 g
UN Number UN2811
Beilstein 2442261
Formula Weight 235.46
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-5-chlorobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.