missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methylphenyl isothiocyanate, 98%, Thermo Scientific™

Product Code. 11337416
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337416 5 g 5g
11327416 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337416 Supplier Thermo Scientific Alfa Aesar Supplier No. L09413.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19241-39-5
Molecular Formula C8H6BrNS
Molecular Weight (g/mol) 228.107
MDL Number MFCD00041040
InChI Key DQXSCOWMTPPFCJ-UHFFFAOYSA-N
Synonym 2-bromo-4-methylphenyl isothiocyanate, 2-bromo-4-methylphenylisothiocyanate, benzene,2-bromo-1-isothiocyanato-4-methyl, acmc-1bo1e, 2-bromo-4-methylisothiocyanatobenzene, #, 2-bromanyl-1-isothiocyanato-4-methyl-benzene
PubChem CID 140506
IUPAC Name 2-bromo-1-isothiocyanato-4-methylbenzene
SMILES CC1=CC(=C(C=C1)N=C=S)Br

Specifications

Density 1.52
Boiling Point 119°C to 120°C (0.5 mmHg)
Odor Irritating
Refractive Index 1.669
Quantity 5 g
UN Number UN2922
Beilstein 2717454
Sensitivity Moisture sensitive
Formula Weight 228.12
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-methylphenyl isothiocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.