Learn More
2-Bromo-4-isopropylaniline, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 51605-97-1 |
| Molecular Formula | C9H12BrN |
| MDL Number | MFCD00051585 |
| Quantity | 25 g |
| SMILES | CC(C)C1=CC(Br)=C(N)C=C1 |
| Molecular Weight (g/mol) | 214.10 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Bromo-4-isopropylaniline |