Learn More
2-Bromo-3’,5’-dibenzyloxyacetphenone, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Bromo-3',5'-dibenzyloxyacetphenone |
| CAS | 28924-18-7 |
| Molecular Formula | C22 H19 Br O3 |
| Quantity | 100 mg |
| Synonym | Terbutaline Impurity 47 |
| InChI Formula | InChI=1S/C22H19BrO3/c23-14-22(24)19-11-20(25-15-17-7-3-1-4-8-17)13-21(12-19)26-16-18-9-5-2-6-10-18/h1-13H,14-16H2 |
| SMILES | BrCC(=O)c1cc(OCc2ccccc2)cc(OCc3ccccc3)c1 |
| IUPAC Name | 1-[3,5-bis(phenylmethoxy)phenyl]-2-bromoethanone |
| Molecular Weight (g/mol) | 411.29 |
| Formula Weight | 410.0518 |