missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-2-methyl-d3-propionic-3,3,3-d3 Acid, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 2-Bromoisobutyric-d6 Acid |
| CAS | 1219795-23-9 |
| Molecular Formula | C4 D6 H Br O2 |
| Quantity | 0.25 g |
| Synonym | 2-Bromo-2-methyl-propanoic-d6 Acid, α-Bromo-isobutyric-d6 Acid, 2-Bromo-2-methyl-propionic-d6 Acid, 2-Bromo-2-methylpropanoic-d6 Acid, 2-Bromo-2-methylpropionic-d6 Acid, 2-Bromoisobutyric-d6 Acid, Bromoisobutyric-d6 Acid, Isobromobutyric-d6 Acid, NSC 41213-d6, α-Bromo-α-methylpropionic-d6 Acid, α-Bromoisobutyric-d6 Acid |
| InChI Formula | InChI=1 S/C4H7BrO2/c1-4(2,5)3(6)7/h1-2H3,(H,6,7)/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])C(Br)(C(=O)O)C([2 H])([2 H])[2 H] |
| IUPAC Name | 2-bromo-3,3,3-trideuterio-2-(trideuteriomethyl)propanoic acid |
| Molecular Weight (g/mol) | 173.04 |
| Formula Weight | 172.0006 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction