Learn More
2-Bromo-1-chloropropane (>90%), TRC
Product Code. 30004233
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
1g
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004233 500 mg 500mg
17943055 2.5 g 2.5g
17974965 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30004233 Supplier Toronto Research Chemicals Supplier No. TRCB680515500MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3017-95-6
Molecular Formula C3 H6 Br Cl
Molecular Weight (g/mol) 157.44
Synonym Propane, 2-bromo-1-chloro-, 2-Bromo-1-chloropropane, (±)-2-Bromo-1-chloropropane, 1-Chloro-2-bromopropane, NSC 8022
IUPAC Name 2-bromo-1-chloropropane
SMILES CC(Br)CCl
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C3H6BrCl/c1-3(4)2-5/h3H,2H2,1H3
Formula Weight 155.9341
Chemical Name or Material 2-Bromo-1-chloropropane