missing translation for 'onlineSavingsMsg'
Learn More

2-(Benzylthio)-4-chlorobenzoic Acid, TRC

Product Code. 30345443
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30345443 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30345443 Supplier Toronto Research Chemicals Supplier No. TRCB315030100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Benzylthio)-4-chlorobenzoic Acid
CAS 40183-35-5
Molecular Formula C14 H11 Cl O2 S
Quantity 100 mg
InChI Formula InChI=1S/C14H11ClO2S/c15-11-6-7-12(14(16)17)13(8-11)18-9-10-4-2-1-3-5-10/h1-8H,9H2,(H,16,17)
SMILES OC(=O)c1ccc(Cl)cc1SCc2ccccc2
IUPAC Name 2-benzylsulfanyl-4-chlorobenzoic acid
Molecular Weight (g/mol) 278.75
Formula Weight 278.0168
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.