missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxypyridine-3-boronic acid pinacol ester, 95%

Product Code. 15457635
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15457635 250 mg 250mg
11421510 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15457635 Supplier Thermo Scientific Alfa Aesar Supplier No. H26928.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1073371-81-9 | C18H22BNO3 | 311.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073371-81-9
Molecular Formula C18H22BNO3
Molecular Weight (g/mol) 311.19
MDL Number MFCD06798265
InChI Key IFMSDMPTVPMGOP-UHFFFAOYSA-N
Synonym 2-benzyloxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-benzyloxypyridine-3-boronic acid pinacol ester, acmc-20aajr, 2-benzyloxy pyridin-3-yl boronic acid pinacol ester, 2-benzyloxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 2-benzyloxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-benzoxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-phenylmethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine
PubChem CID 44755162
IUPAC Name 2-phenylmethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1(C)OB(OC1(C)C)C1=C(OCC2=CC=CC=C2)N=CC=C1

Specifications

Quantity 250 mg
Formula Weight 311.19
Percent Purity 95%
Chemical Name or Material 2-Benzyloxypyridine-3-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.