missing translation for 'onlineSavingsMsg'
Learn More

2-(Benzyloxy)-1-ethanamine hydrochloride, ≥90%, Thermo Scientific™

Product Code. 10462813
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10462813

Brand: Thermo Scientific Maybridge MO07103EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 10578-75-3
Molecular Formula C9H14ClNO
Molecular Weight (g/mol) 187.667
MDL Number MFCD06411550
InChI Key AFQMWBUNDONXED-UHFFFAOYSA-N
Synonym 2-benzyloxy ethanamine hydrochloride, 2-benzyloxy-1-ethanamine hydrochloride, 2-benzyloxyethylamine hydrochloride, aminoethylbenzyl ether hydrochloride, 2-phenylmethoxy-ethylamine hydrochloride, 2-benzyloxy-1-ethanamine, hcl, 2-benzyloxy ethylamine hydrochloride, 2-benzyloxy ethyl amine hydrochloride, ethanamine, 2-phenylmethoxy-, hydrochloride, 2-phenylmethoxy ethylamine, chloride
PubChem CID 2794786
IUPAC Name 2-phenylmethoxyethanamine;hydrochloride
SMILES C1=CC=C(C=C1)COCCN.Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 187.67
Percent Purity 97%
Chemical Name or Material 2-(Benzyloxy)-1-ethanamine hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.