missing translation for 'onlineSavingsMsg'
Learn More

2-Benzylamino-2-phenylbutanol, TRC

Product Code. 30335725
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335725 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335725 Supplier Toronto Research Chemicals Supplier No. TRCB225990100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Benzylamino-2-phenylbutanol
CAS 1178412-63-9
Molecular Formula C17H21NO
Quantity 100 mg
Synonym N-Benzyl 1-Phenyl-1-hydroxymetbl-1-propanamine, beta-Ethyl-beta-[(phenylmethyl)amino]benzeneethanol
InChI Formula InChI=1S/C17H21NO/c1-2-17(14-19,16-11-7-4-8-12-16)18-13-15-9-5-3-6-10-15/h3-12,18-19H,2,13-14H2,1H3
SMILES CCC(CO)(NCc1ccccc1)c2ccccc2
IUPAC Name 2-(benzylamino)-2-phenylbutan-1-ol
Molecular Weight (g/mol) 255.35
Formula Weight 255.1623
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.