Learn More
2-(Benzyl(methyl)amino)ethanol, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 101-98-4 |
| Molecular Formula | C10H15NO |
| MDL Number | MFCD00002847 |
| Quantity | 1 kg |
| SMILES | CN(CCO)CC1=CC=CC=C1 |
| Molecular Weight (g/mol) | 165.23 |
| Percent Purity | 95% |
| Chemical Name or Material | 2-(Benzyl(methyl)amino)ethanol |