missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyl-D-proline hydrochloride, 95%, Thermo Scientific Chemicals

Product Code. 15458507
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
missing translation for 'unitSize'
1g
250mg
This item is not returnable. View return policy

Product Code. 15458507

missing translation for 'mfr': Thermo Scientific Alfa Aesar H52019.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1111130-04-1
Molecular Formula C12H16ClNO2
Molecular Weight (g/mol) 241.72
MDL Number MFCD06659152
InChI Key SPHJFGQFCBZKRU-UHFFFAOYNA-N
Synonym s-2-benzylpyrrolidine-2-carboxylic acid hydrochloride, s-alpha-benzylproline hcl, 2-benzyl-d-proline hydrochloride, 2s-2-benzylpyrrolidine-2-carboxylic acid hydrochloride, d-proline,2-phenylmethyl, s-2-benzyl-d-proline hydrochloride, s-alpha-benzyl-proline hydrochloride, 2-benzyl-d-proline-hydrogen chloride 1/1
PubChem CID 45026885
SMILES Cl.OC(=O)C1(CC2=CC=CC=C2)CCCN1

Specifications

IUPAC Name 2-benzylpyrrolidine-2-carboxylic acid hydrochloride
Quantity 250 mg
Formula Weight 241.72
Optical Rotation +33° (c=1 in 1N HCl)
Percent Purity 95%
Chemical Name or Material 2-Benzyl-D-proline hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.