Learn More
2-Benzyl-1,3-propanediol, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Benzyl-1,3-propanediol |
| CAS | 2612-30-8 |
| Molecular Formula | C10H14O2 |
| Quantity | 1 g |
| Synonym | 2-(Phenylmethyl)-1,3-propanediol |
| InChI Formula | InChI=1S/C10H14O2/c11-7-10(8-12)6-9-4-2-1-3-5-9/h1-5,10-12H,6-8H2 |
| SMILES | OCC(CO)CC1=CC=CC=C1 |
| IUPAC Name | 2-Benzyl-1,3-propanediol |
| Molecular Weight (g/mol) | 166.22 |
| Formula Weight | 166.1 |