Learn More
2-Aminopyridine-6-methanol, 97%
Description
4-Aminopyridine (4-AP), a well-known potassium channel blocker, has been shown to enhance conduction in injured and demyelinated axons. Aminopyridine has a narrow therapeutic range in clinical application. In the current study, we investigated whether AP-MeOH can enhance impulse conduction in an animal model of MS.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Slightly soluble.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 87°C to 92°C |
| Quantity | 5 g |
| Solubility Information | Slightly soluble. |
| Formula Weight | 124.14 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Aminopyridine-6-methanol |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.