Learn More
2-Aminoethyl Methanethiosulfonate Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Aminoethyl Methanethiosulfonate, Hydrochloride |
| CAS | 37597-96-9 |
| Molecular Formula | C3 H9 N O2 S2 . Cl H |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C3H9NO2S2.ClH/c1-8(5,6)7-3-2-4;/h2-4H2,1H3;1H |
| SMILES | Cl.CS(=O)(=O)SCCN |
| IUPAC Name | 2-methylsulfonylsulfanylethanamine;hydrochloride |
| Molecular Weight (g/mol) | 191.7 |
| Formula Weight | 190.9841 |