Learn More
2-Amino-n-(tert-butyl)benzenesulfonamide, TRC
Product Code. 30340258
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30340258 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30340258 Supplier Toronto Research Chemicals Supplier No. TRCA639815100MG
 Restricted Item
Estimated Shipment: 01-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Amino-n-(tert-butyl)benzenesulfonamide
CAS 954268-81-6
Molecular Formula C10H16N2O2S
Quantity 100 mg
InChI Formula InChI=1S/C10H16N2O2S/c1-10(2,3)12-15(13,14)9-7-5-4-6-8(9)11/h4-7,12H,11H2,1-3H3
SMILES CC(C)(C)NS(=O)(=O)C1=CC=CC=C1N
IUPAC Name 2-amino-N-tert-butylbenzenesulfonamide
Molecular Weight (g/mol) 228.31
Formula Weight 228.0932