Learn More
2-Amino-N-isopropylbenzamide, TRC
Specifications
Specifications
| Chemical Name or Material | Anthranilic acid isopropylamide |
| CAS | 30391-89-0 |
| Molecular Formula | C10 H14 N2 O |
| Quantity | 1 g |
| Synonym | Benzamide, o-amino-N-isopropyl- (8CI), 2-Amino-N-(1-methylethyl)benzamide, 2-Amino-N-isopropylbenzamide, Anthranilic acid isopropylamide, N-Isopropylanthranilamide, NSC 88050 |
| InChI Formula | InChI=1S/C10H14N2O/c1-7(2)12-10(13)8-5-3-4-6-9(8)11/h3-7H,11H2,1-2H3,(H,12,13) |
| SMILES | CC(C)NC(=O)c1ccccc1N |
| IUPAC Name | 2-amino-N-propan-2-ylbenzamide |
| Molecular Weight (g/mol) | 178.23 |
| Formula Weight | 178.1106 |