Learn More
2-Amino-6-chlorobenzonitrile, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 2-Amino-6-chlorobenzonitrile |
| CAS | 6575-11-7 |
| Molecular Formula | C7H5ClN2 |
| Quantity | 10 g |
| SMILES | Nc1cccc(Cl)c1C#N |
| Molecular Weight (g/mol) | 152.58 |
| Formula Weight | 152.0141 |