Learn More
2-Amino-6-chloro-4-pyrimidinol, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Amino-6-chloro-4-pyrimidinol |
| CAS | 1194-21-4 |
| Molecular Formula | C4H4ClN3O |
| Quantity | 1 g |
| Synonym | 2-Amino-4-chloro-6-hydroxypyrimidine, 2-Amino-4-chloropyrimidine-6-one, 2-Amino-4-hydroxy-6-chloropyrimidine, 2-Amino-6-chloro-3H-pyrimidin-4-one, 2-Amino-6-chloro-4(1H)-pyrimidinone, 2-Amino-6-chloro-4(3H)-pyrimidinone |
| InChI Formula | InChI=1S/C4H4ClN3O/c5-2-1-3(9)8-4(6)7-2/h1H,(H3,6,7,8,9) |
| SMILES | ClC(NC(N)=N1)=CC1=O |
| IUPAC Name | 2-amino-6-chloropyrimidin-4(1H)-one |
| Molecular Weight (g/mol) | 145.55 |
| Formula Weight | 145 |