Learn More
2-Amino-6-chloro-4-pyrimidinol, TRC
Product Code. 30327178
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30327178 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30327178 Supplier Toronto Research Chemicals Supplier No. TRCA603195250MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Amino-6-chloro-4-pyrimidinol
CAS 1194-21-4
Molecular Formula C4H4ClN3O
Quantity 250 mg
Synonym 2-Amino-4-chloro-6-hydroxypyrimidine, 2-Amino-4-chloropyrimidine-6-one, 2-Amino-4-hydroxy-6-chloropyrimidine, 2-Amino-6-chloro-3H-pyrimidin-4-one, 2-Amino-6-chloro-4(1H)-pyrimidinone, 2-Amino-6-chloro-4(3H)-pyrimidinone
InChI Formula InChI=1S/C4H4ClN3O/c5-2-1-3(9)8-4(6)7-2/h1H,(H3,6,7,8,9)
SMILES ClC(NC(N)=N1)=CC1=O
IUPAC Name 2-amino-6-chloropyrimidin-4(1H)-one
Molecular Weight (g/mol) 145.55
Formula Weight 145