missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-Ethylphenol Hydrochloride, TRC

Product Code. 30335889
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335889 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335889 Supplier Toronto Research Chemicals Supplier No. TRCA898120100MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Amino-5-Ethylphenol Hydrochloride
CAS 149861-22-3
Molecular Formula C8H11NO . HCl
Quantity 100 mg
InChI Formula InChI=1S/C8H11NO.ClH/c1-2-6-3-4-7(9)8(10)5-6;/h3-5,10H,2,9H2,1H3;1H
SMILES Cl.CCc1ccc(N)c(O)c1
IUPAC Name 2-amino-5-ethylphenol;hydrochloride
Molecular Weight (g/mol) 173.64
Formula Weight 173.0607
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.