Learn More
2-Amino-5-chlorobenzyl alcohol, 98%
Product Code. 15483947
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15483947 5 g 5g
15493947 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15483947 Supplier Thermo Scientific Alfa Aesar Supplier No. H30531.06
Request Bulk or Custom Format
CAS: 37585-25-4 | C7H8ClNO | 157.597 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 37585-25-4
Molecular Formula C7H8ClNO
Molecular Weight (g/mol) 157.597
MDL Number MFCD00075163
InChI Key CLKBZWDZDVOIGJ-UHFFFAOYSA-N
PubChem CID 586754
IUPAC Name (2-amino-5-chlorophenyl)methanol
SMILES C1=CC(=C(C=C1Cl)CO)N
Specifications
Melting Point 107°C to 109°C
Quantity 5 g
Sensitivity Air Sensitive
Synonym 2-amino-5-chlorophenyl methanol, 2-amino-5-chlorobenzyl alcohol, 2-amino-5-chloro-phenyl-methanol, benzenemethanol, 2-amino-5-chloro, acmc-20ao4o, 2-amino-5-chlorobenzenemethanol, 2-hydroxymethyl-4-chloroaniline, benzenemethanol,2-amino-5-chloro, 2-amino-5-chlorophenyl methanol #
Formula Weight 157.6
Percent Purity 98%
Chemical Name or Material 2-Amino-5-chlorobenzyl alcohol