missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4-thiazoleacetic Acid-13C,15N2, TRC
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
Specifications
Specifications
| Chemical Name or Material | 2-Amino-4-thiazoleacetic Acid-13C,15N2 |
| Molecular Formula | 13C C4 H6 15N2 O2 S |
| Quantity | 10 mg |
| Synonym | (2-Amino-1,3-thiazol-4-yl)acetic Acid-13C,15N2, (2-Amino-4-thiazoyl)acetic Acid-13C,15N2, 2-(2-Amino-4-thiazolyl)acetic Acid-13C,15N2, 2-Amino-4-thiazolylacetic Acid-13C,15N2, 2-Aminothiazole-4-acetic Acid-13C,15N2, SQ 272-13C,15N2 |
| InChI Formula | InChI=1S/C5H6N2O2S/c6-5-7-3(2-10-5)1-4(8)9/h2H,1H2,(H2,6,7)(H,8,9)/i5+1,6+1,7+1 |
| SMILES | [15NH2][13c]1[15n]c(CC(=O)O)cs1 |
| IUPAC Name | 2-(2-(15N)azanyl-(213C,315N)1,3-thiazol-4-yl)acetic acid |
| Molecular Weight (g/mol) | 161.158 |
| Formula Weight | 161.012 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction