missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-thiazoleacetic Acid-13C,15N2, TRC

Product Code. 30340870
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 30340870

Brand: Toronto Research Chemicals TRCA63048625MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Amino-4-thiazoleacetic Acid-13C,15N2
Molecular Formula 13C C4 H6 15N2 O2 S
Quantity 25 mg
Synonym (2-Amino-1,3-thiazol-4-yl)acetic Acid-13C,15N2, (2-Amino-4-thiazoyl)acetic Acid-13C,15N2, 2-(2-Amino-4-thiazolyl)acetic Acid-13C,15N2, 2-Amino-4-thiazolylacetic Acid-13C,15N2, 2-Aminothiazole-4-acetic Acid-13C,15N2, SQ 272-13C,15N2
InChI Formula InChI=1S/C5H6N2O2S/c6-5-7-3(2-10-5)1-4(8)9/h2H,1H2,(H2,6,7)(H,8,9)/i5+1,6+1,7+1
SMILES [15NH2][13c]1[15n]c(CC(=O)O)cs1
IUPAC Name 2-(2-(15N)azanyl-(213C,315N)1,3-thiazol-4-yl)acetic acid
Molecular Weight (g/mol) 161.158
Formula Weight 161.012
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.