missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Amino-4-(methoxycarbonyl)benzeneboronic acid pinacol ester hydrochloride, 95%

Product Code. 15475245
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15475245 1 g 1g
15485245 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15475245 Supplier Alfa Aesar™ Supplier No. H53220.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy
Applications
2-Amino-4-(methoxycarbonyl)benzeneboronic acid pinacol ester hydrochloride is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850567-49-6
Molecular Formula C14H21BClNO4
Molecular Weight (g/mol) 313.585
MDL Number MFCD06659918
InChI Key HEBCDUUNEDAXAA-UHFFFAOYSA-N
Synonym methyl 3-amino-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate hydrochloride, 2-amino-4-methoxycarbonylphenylboronic acid, pinacol ester, hcl, 2-amino-4-methoxycarbonylphenyl boronic acid pinacol ester hydrochloride, methyl 3-amino-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate hydrochloride, methyl 3-amino-4-boronobenzoate, pinacol ester, hcl, 2-amino-4-methoxycarbonyl benzeneboronic acid pinacol ester hydrochloride, 2-amino-4-methoxycarbonylphenylboronic acid,pinacol, methyl 3-amino-4-boronobenzoate pinacol ester hydrochloride, 2-amino-4-methoxycarbonyl benzeneboronic acid, pinacol ester hydrochloride, 2-amino-4-methoxycarbonylphenylboronic acid pinacol ester hydrochloride
PubChem CID 44119628
IUPAC Name methyl 3-amino-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate;hydrochloride
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C(=O)OC)N.Cl

Specifications

Melting Point 92°C to 94°C
Quantity 1 g
Formula Weight 313.59
Percent Purity 95%
Chemical Name or Material 2-Amino-4-(methoxycarbonyl)benzeneboronic acid pinacol ester hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.