missing translation for 'onlineSavingsMsg'
Learn More

2-amino-3-pyridinecarboxaldehyde 97%, Thermo Scientific™

Product Code. 10311213 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10311213 5 g Glass bottle 5g
10004192 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10311213 Supplier Thermo Scientific Acros Supplier No. 380610050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7521-41-7
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.127
MDL Number MFCD01830382
InChI Key NXMFJCRMSDRXLD-UHFFFAOYSA-N
Synonym 2-aminonicotinaldehyde, 2-amino-3-pyridinecarboxaldehyde, 2-amino-3-formylpyridine, 3-pyridinecarboxaldehyde, 2-amino, 2-amino-pyridine-3-carbaldehyde, 2-aminopyridine-3-carboxaldehyde, 2-amino-3-pyridine carboxaldehyde, 2-amino-pyridin-3-carbaldehyde, pubchem1262, 2-amino nicotinaldehyde
PubChem CID 737633
IUPAC Name 2-aminopyridine-3-carbaldehyde
SMILES C1=CC(=C(N=C1)N)C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 95°C to 102°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 122.13
Percent Purity ≥96.0%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 2-Amino-3-pyridinecarboxaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.