Learn More
2-Amino-3-pyridinecarbonitrile, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 2-Amino-3-pyridinecarbonitrile |
| CAS | 24517-64-4 |
| Molecular Formula | C6H5N3 |
| Quantity | 5 g |
| Synonym | 2-Aminonicotinonitrile, 2-Amino-3-cyanopyridine, 2-Aminonicotinonitrile, 3-Cyanopyridin-2-amine, 3-Cyanopyridin-2-ylamine |
| InChI Formula | InChI=1S/C6H5N3/c7-4-5-2-1-3-9-6(5)8/h1-3H,(H2,8,9) |
| SMILES | NC1=NC=CC=C1C#N |
| IUPAC Name | 2-aminonicotinonitrile |
| Molecular Weight (g/mol) | 119.13 |
| Formula Weight | 119.05 |