missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-diiodobenzoic acid, 98+%, Thermo Scientific™

Product Code. 11448186
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448186 5 g 5g
11458186 25 g 25g
11468186 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448186 Supplier Thermo Scientific Alfa Aesar Supplier No. A15308.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-86-9
Molecular Formula C7H5I2NO2
Molecular Weight (g/mol) 388.931
MDL Number MFCD00007682
InChI Key RFIBDMCPIREZKC-UHFFFAOYSA-N
Synonym 3,5-diiodoanthranilic acid, benzoic acid, 2-amino-3,5-diiodo, anthranilic acid, 3,5-diiodo, 2-amino-3,5-diiodobenzoicacid, acmc-209mnv, 3,5-diiodoanthranilicacid, 3,5-diiodoanthanilic acid, anthranilic acid,5-diiodo, 4-14-00-01085 beilstein handbook reference, 2-amino-3,5-diiodo-benzoic acid
PubChem CID 69116
IUPAC Name 2-amino-3,5-diiodobenzoic acid
SMILES C1=C(C=C(C(=C1I)N)C(=O)O)I

Specifications

Melting Point ∼240°C (decomposition)
Quantity 5 g
Beilstein 779429
Sensitivity Light sensitive
Formula Weight 388.93
Percent Purity ≥98%
Chemical Name or Material 2-Amino-3,5-diiodobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.