Learn More
2-Amino-3,5-dichlorobenzoic acid, 97%
Product Code. p-7023422
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Product Code. 11408806
Brand: Thermo Scientific Alfa Aesar A16969.09
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
CAS: 2789-92-6 | C7H5Cl2NO2 | 206.02 g/mol
2-Amino-3,5-dichlorobenzoic acid, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-3,5-dichlorobenzoic acid, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
This material is stable if stored under proper conditions. Avoid excessive heat and light. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2789-92-6
Molecular Formula C7H5Cl2NO2
Molecular Weight (g/mol) 206.02
MDL Number MFCD00017088
InChI Key KTHTXLUIEAIGCD-UHFFFAOYSA-N
Synonym 3,5-dichloroanthranilic acid, benzoic acid, 2-amino-3,5-dichloro, anthranilic acid, 3,5-dichloro, 3,5-dichloro-2-aminobenzoic acid, 2-amino-3,5-dichloro-benzoic acid, timtec-bb sbb003493, 2-amino-3,5-dichlorobenzoicacid, acmc-209gzl, rarechem al bo 0770, buttpark 49\07-46
PubChem CID 76036
IUPAC Name 2-amino-3,5-dichlorobenzoic acid
SMILES NC1=C(Cl)C=C(Cl)C=C1C(O)=O
Specifications
Melting Point 225°C to 227°C
Quantity 10 g
Beilstein 779100
Solubility Information Insoluble in water.
Formula Weight 206.03
Percent Purity 97%
Chemical Name or Material 2-Amino-3,5-dichlorobenzoic acid