missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-methylpropanamide Hydrochloride, TRC

Product Code. 30318516
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30318516 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318516 Supplier Toronto Research Chemicals Supplier No. TRCA010213100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Amino-2-methylpropanamide Hydrochloride
CAS 17704-74-4
Molecular Formula C4H10N2O . HCl
Quantity 100 mg
InChI Formula InChI=1S/C4H10N2O.ClH/c1-4(2,6)3(5)7;/h6H2,1-2H3,(H2,5,7);1H
SMILES Cl.CC(C)(N)C(=O)N
IUPAC Name 2-amino-2-methylpropanamide;hydrochloride
Molecular Weight (g/mol) 138.59
Formula Weight 138.056
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.