Learn More
2-Amino-1-cyclopentene-1-carbonitrile, TRC
Product Code. 30132934
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132934 10 g 10g
30009469 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30132934 Supplier Toronto Research Chemicals Supplier No. TRCA60408010G
 Restricted Item
Estimated Shipment: 03-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2941-23-3
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
Synonym 1-Amino-2-cyano-1,2-cyclopentene, 1-Amino-2-cyano-1-cyclopentene, 1-Amino-2-cyanocyclopentene, 2-Amino-1-cyano-1-cyclopentene, 2-Amino-1-cyclopentene-1-carbonitrile, 2-Aminocyclopent-1-enecarbonitrile, 2-Cyanocyclopenten-1-ylamine, NSC 146852
IUPAC Name 2-aminocyclopentene-1-carbonitrile
SMILES NC1=C(CCC1)C#N
Specifications
Quantity 10 g
InChI Formula InChI=1S/C6H8N2/c7-4-5-2-1-3-6(5)8/h1-3,8H2
Formula Weight 108.0687
Chemical Name or Material 2-Amino-1-cyclopentene-1-carbonitrile