Learn More
2-Amino-1-(4-morpholinyl)-ethanone Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Amino-1-(4-morpholinyl)-ethanone Hydrochloride |
| CAS | 24152-96-3 |
| Molecular Formula | C6H12N2O2.HCl |
| Quantity | 25 mg |
| Synonym | 2-Amino-1-(4-morpholinyl)-ethanone Hydrochloride |
| InChI Formula | InChI=1S/C6H12N2O2.ClH/c7-5-6(9)8-1-3-10-4-2-8;/h1-5,7H2;1H |
| SMILES | NCC(N1CCOCC1)=O.[H]Cl |
| IUPAC Name | 2-Amino-1-(4-morpholinyl)-ethanone Hydrochloride |
| Molecular Weight (g/mol) | 180.63 |
| Formula Weight | 180.07 |