Learn More
Julabo™ 2-Adapters for Cryo Compact Circulator
Product Code. p-8013515
Click to view available options
Connection Type:
M16 x 1 Female to 1/2in. Tube Male
M16 x 1 Female to NPT 1/4in. Female
M16 x 1 Female to NPT 3/8in. Female
Unit Size:
Pack of 2
Product Code. 11934698
Brand: Julabo™ 8890005
Only null left
Specifications
For Use With (Equipment) Suitable for CF30, CF31, CF40, CF41 Cryo-Compact Circulator
Type 2-Adapters
Connection Type M16 x 1 Female to NPT 1/4in. Female