Learn More
Julabo™ 2-Adapters for Cryo Compact Circulator
Click to view available options
Connection Type:
M16 x 1 Female to 1/2in. Tube Male
M16 x 1 Female to NPT 1/4in. Female
M16 x 1 Female to NPT 3/8in. Female
Unit Size:
Pack of 2
Specifications
Specifications
| For Use With (Equipment) | Suitable for CF30, CF31, CF40, CF41 Cryo-Compact Circulator |
| Type | 2-Adapters |
| Connection Type | M16 x 1 Female to NPT 1/4in. Female |