missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Acetamidoacrylic Acid, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | 2-Acetamidoacrylic Acid |
| CAS | 5429-56-1 |
| Molecular Formula | C5H7NO3 |
| Quantity | 250 mg |
| Synonym | 2-(Acetylamino)-2-propenoic Acid, 2-(Acetylamino)acrylic Acid, Acetyldehydroalanine, N-Acetyl-alpha-aminoacrylic Acid, NSC 14171 |
| InChI Formula | InChI=1S/C5H7NO3/c1-3(5(8)9)6-4(2)7/h1H2,2H3,(H,6,7)(H,8,9) |
| SMILES | CC(=O)NC(=C)C(=O)O |
| IUPAC Name | 2-acetamidoprop-2-enoic acid |
| Molecular Weight (g/mol) | 129.11 |
| Formula Weight | 129.0426 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction