missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamidoacrylic acid, 97+%

Product Code. 11361367
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11361367

Brand: Thermo Scientific Alfa Aesar L01649.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5429-56-1 | C5H7NO3 | 129.115 g/mol

2-Acetamidoacrylic acid is used as an antioxidant as well as a monomer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetamidoacrylic acid is used as an antioxidant as well as a monomer.

Solubility
Soluble in dimethyl sulfoxide. Slightly soluble in water and methanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5429-56-1
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.115
MDL Number MFCD00004257
InChI Key UFDFFEMHDKXMBG-UHFFFAOYSA-N
Synonym 2-acetamidoacrylic acid, 2-propenoic acid, 2-acetylamino, acrylic acid, 2-acetamido, 2-acetamidoacrylicacid, acetyldehydroalanine, n-acetyldehydroalanine, 2-acetylamino acrylic acid, .alpha.-acetamidoacrylic acid, 2-acetylamino-2-propenoic acid, 2-acetylamino prop-2-enoic acid
PubChem CID 79482
IUPAC Name 2-acetamidoprop-2-enoic acid
SMILES CC(=O)NC(=C)C(=O)O

Specifications

Melting Point ∽190°C (decomposition)
Quantity 25 g
Beilstein 1812032
Solubility Information Soluble in dimethyl sulfoxide. Slightly soluble in water and methanol.
Formula Weight 129.12
Percent Purity 99%
Chemical Name or Material 2-Acetamidoacrylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.