Learn More
2-Acetamidoacrylic acid, 97+%
Product Code. 11341367
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11341367
Brand: Thermo Scientific Alfa Aesar L01649.03
Request Bulk or Custom Format
CAS: 5429-56-1 | C5H7NO3 | 129.115 g/mol
2-Acetamidoacrylic acid is used as an antioxidant as well as a monomer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Acetamidoacrylic acid is used as an antioxidant as well as a monomer.
Solubility
Soluble in dimethyl sulfoxide. Slightly soluble in water and methanol.
Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5429-56-1
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.115
MDL Number MFCD00004257
InChI Key UFDFFEMHDKXMBG-UHFFFAOYSA-N
Synonym 2-acetamidoacrylic acid, 2-propenoic acid, 2-acetylamino, acrylic acid, 2-acetamido, 2-acetamidoacrylicacid, acetyldehydroalanine, n-acetyldehydroalanine, 2-acetylamino acrylic acid, .alpha.-acetamidoacrylic acid, 2-acetylamino-2-propenoic acid, 2-acetylamino prop-2-enoic acid
PubChem CID 79482
IUPAC Name 2-acetamidoprop-2-enoic acid
SMILES CC(=O)NC(=C)C(=O)O
Specifications
Melting Point ∽190°C (decomposition)
Quantity 1 g
Beilstein 1812032
Solubility Information Soluble in dimethyl sulfoxide. Slightly soluble in water and methanol.
Formula Weight 129.12
Percent Purity 99%
Chemical Name or Material 2-Acetamidoacrylic acid