Learn More
2-Acetamido-4,6-O-benzylidene-2-deoxy-Beta-D-glucopyranosyl Azide, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Acetamido-4,6-O-benzylidene-2-deoxy-beta-D-glucopyranosyl Azide |
| CAS | 168397-51-1 |
| Molecular Formula | C15H18N4O5 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C15H18N4O5/c1-8(20)17-11-12(21)13-10(23-14(11)18-19-16)7-22-15(24-13)9-5-3-2-4-6-9/h2-6,10-15,21H,7H2,1H3,(H,17,20) |
| SMILES | CC(=O)N[C@@H]1[C@@H](O)[C@@H]2O[C@@H](OC[C@H]2O[C@H]1N=[N+]=[N-])c3ccccc3 |
| IUPAC Name | N-(6-azido-8-hydroxy-2-phenyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-7-yl)acetamide |
| Molecular Weight (g/mol) | 334.33 |
| Formula Weight | 334.1277 |